missing translation for 'onlineSavingsMsg'
Learn More

Exosome Component 9 Antibody (1090209) [mFluor Violet 610 SE], Novus Biologicals™

Product Code. 30762735 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30762735 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30762735 Supplier Novus Biologicals Supplier No. FAB11616MFV610

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Exosome Component 9 Monoclonal antibody specifically detects Exosome Component 9 in Human samples. It is validated for Immunohistochemistry,Immunocytochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen Exosome Component 9
Applications Immunohistochemistry, Immunocytochemistry
Classification Monoclonal
Clone 1090209
Conjugate mFluor Violet 610 SE
Formulation 50mM Sodium Borate
Gene Alias Autoantigen PM/Scl 1, EC 3.1.13, exosome complex component RRP45, exosome complex exonuclease RRP45, P5, p6, P75 polymyositis-scleroderma overlap syndrome associated autoantigen, PCH1D, PM/Scl-75, PMSCL autoantigen, 75kD, PMSCL1, Polymyositis/scleroderma autoantigen 1, polymyositis/scleroderma autoantigen 1 (75kD), polymyositis/scleroderma autoantigen 1, 75kDa, RRP45, Rrp45p
Host Species Mouse
Immunogen Synthetic Peptide, Accession # Q06265
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5393
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.