missing translation for 'onlineSavingsMsg'
Learn More

Exosome component 5 Antibody, Alexa Fluor™ 532, Novus Biologicals™

Product Code. 18429715 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18429715 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18429715 Supplier Novus Biologicals Supplier No. NBP222241AF532

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Exosome component 5 Polyclonal antibody specifically detects Exosome component 5 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Exosome component 5
Applications Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 532
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias CML28, exosome complex exonuclease RRP46, exosome component 5Chronic myelogenous leukemia tumor antigen 28, exosome component Rrp46, hRrp46p, MGC12901, p12BMGC111224, RRP41B, Rrp46p, RRP46Ribosomal RNA-processing protein 46
Host Species Rabbit
Immunogen The immunogen this antibody was made to, maps to a region between residue 1 and 50 of human Exosome Component 5 using the numbering given in entry NP_064543.3 (GeneID 56915).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline DNA replication Transcription Translation and Splicing, Epigenetics
Primary or Secondary Primary
Gene ID (Entrez) 56915
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.