missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Exosome component 1 Antibody (OTI1H9), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71895F
This item is not returnable.
View return policy
Description
Exosome component 1 Monoclonal specifically detects Exosome component 1 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry.
Specifications
| Exosome component 1 | |
| Monoclonal | |
| FITC | |
| PBS with 0.05% Sodium Azide | |
| EXOSC1 | |
| Full length recombinant protein of human Exosome component 1 (NP_057130) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Immunohistochemistry | |
| OTI1H9 | |
| Immunohistochemistry | |
| CGI-108,3′-5′ exoribonuclease CSL4 homolog, CSL4 exosomal core protein homolog, Csl4p, CSL4p13, exosomal core protein CSL4, exosome component 1hCsl4p, homolog of yeast exosomal core protein CSL4, SKI4, Ski4p | |
| Mouse | |
| Affinity Purified | |
| Immunology | |
| 51013.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction