Learn More
Invitrogen™ EXOSC8 Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA5100245
Product Code. 16606734
  • £448.00 / 100µL
Description
Antibody detects endogenous levels of total EXOSC8.
This gene encodes a 3'-5' exoribonuclease that specifically interacts with mRNAs containing AU-rich elements. The encoded protein is part of the exosome complex that is important for the degradation of numerous RNA species. A pseudogene of this gene is found on chromosome 6.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
EXOSC8
Polyclonal
Unconjugated
EXOSC8
2310032N20Rik; bA421P11.3; CBP-interacting protein 3; CIP3; EAP2; EXOSC8; Exosome complex component RRP43; exosome complex exonuclease RRP43; exosome component 8; mKIAA4013; OIP2; OIP-2; Opa interacting protein 2; Opa-interacting protein 2; p9; PCH1C; Ribosomal RNA-processing protein 43; RP11-421P11.3; RRP43; Rrp43p
Rabbit
Affinity chromatography
RUO
11340, 295050, 69639
-20°C
Liquid
Western Blot
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q96B26, Q9D753
EXOSC8
A synthesized peptide derived from human EXOSC8(Accession Q96B26), corresponding to amino acid residues K63-I113.
100 μL
Primary
Human, Mouse, Rat