missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ EXOSC8 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA583471
This item is not returnable.
View return policy
Description
Immunogen sequence: VEPLEYYRRF LKENCRPDGR ELGEFRTTTV NIGSISTADG SALVKLGNTT VICGVKAEFA APSTDAPDKG YVVPNVDLPP LCSSRFR Highest antigen sequence indentity to the following orthologs: Rat - 60%, Mouse - 95%.
This gene encodes a 3'-5' exoribonuclease that specifically interacts with mRNAs containing AU-rich elements. The encoded protein is part of the exosome complex that is important for the degradation of numerous RNA species. A pseudogene of this gene is found on chromosome 6.
Specifications
| EXOSC8 | |
| Polyclonal | |
| Unconjugated | |
| EXOSC8 | |
| 2310032N20Rik; bA421P11.3; CBP-interacting protein 3; CIP3; EAP2; EXOSC8; Exosome complex component RRP43; exosome complex exonuclease RRP43; exosome component 8; mKIAA4013; OIP2; OIP-2; Opa interacting protein 2; Opa-interacting protein 2; p9; PCH1C; Ribosomal RNA-processing protein 43; RP11-421P11.3; RRP43; Rrp43p | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 11340 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96B26 | |
| EXOSC8 | |
| Recombinant protein corresponding to Human EXOSC8. Recombinant protein control fragment (Product #RP-97933). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction