missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EXOSC5 Rabbit anti-Human, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15627-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a tumor suppressor protein that can induce cell growth arrest and apoptosis. The encoded protein is a nuclear protein that physically interacts with the tumor suppressor protein TP53 and is a component of the p53 signaling pathway. Reduced expression and rearrangement of this gene have been detected in various cancers. Multiple alternatively spliced transcript variants encoding distinct isoforms have been reported.Specifications
| EXOSC5 | |
| Polyclonal | |
| Unconjugated | |
| EXOSC5 | |
| CML28, EXOSC5, exosome component 5, hRrp46p, p12B, RRP41B, RRP46, Rrp46p | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 308441, 56915 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.19 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9NQT4 | |
| Exosc5 | |
| EXOSC5 Fusion Protein Ag8098 | |
| 150 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction