Learn More
Invitrogen™ EXOSC5 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5109700
Description
Immunogen sequence: MEEETHTDAK IRAENGTGSS PRGPGCSLRH FACEQNLLSR PDGSASFLQG DTSVLAGVYG PAEVKVSKEI FNKATLEVIL RPKIGLPGVA EKSRERLIRN TCEAVVLGTL HPRTSITVVL QVVSDAGSLL ACCLNAACMA LVDAGVPMRA LFCGVACALD SDGTLVLDPT SKQEKEARAV LTFALDSVER KLLMSSTKGL YSDTELQQCL AAAQAASQHV FRFYRESLQR RYSKS.
Non-catalytic component of the RNA exosome complex which has 3'->5' exoribonuclease activity and participates in a multitude of cellular RNA processing and degradation events. In the nucleus, the RNA exosome complex is involved in proper maturation of stable RNA species such as rRNA, snRNA and snoRNA, in the elimination of RNA processing by-products and non-coding 'pervasive' transcripts, such as antisense RNA species and promoter-upstream transcripts (PROMPTs), and of mRNAs with processing defects, thereby limiting or excluding their export to the cytoplasm. The RNA exosome may be involved in Ig class switch recombination (CSR) and/or Ig variable region somatic hypermutation (SHM) by targeting AICDA deamination activity to transcribed dsDNA substrates. In the cytoplasm, the RNA exosome complex is involved in general mRNA turnover and specifically degrades inherently unstable mRNAs containing AU-rich elements (AREs) within their 3' untranslated regions, and in RNA surveillance pathways, preventing translation of aberrant mRNAs. It seems to be involved in degradation of histone mRNA. The catalytic inactive RNA exosome core complex of 9 subunits (Exo-9) is proposed to play a pivotal role in the binding and presentation of RNA for ribonucleolysis, and to serve as a scaffold for the association with catalytic subunits and accessory proteins or complexes. [UniProt].
Specifications
| EXOSC5 | |
| Polyclonal | |
| Unconjugated | |
| EXOSC5 | |
| Chronic myelogenous leukemia tumor antigen 28; CML28; D7Wsu180e; EXOSC5; Exosome complex component RRP46; exosome complex exonuclease RRP46; exosome complex exonuclease RRP46; exosome complex component RRP46; exosome component 5; exosome component Rrp46; hRrp46p; p12B; Ribosomal RNA-processing protein 46; RRP41B; RRP46; Rrp46p; si:ch211-194n6.2; Unknown (protein for MGC:153707); zgc:153707 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 308441 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.01 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| 0 | |
| EXOSC5 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-235 of human EXOSC5 (NP_064543.3). | |
| 100 μL | |
| Primary | |
| Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.