missing translation for 'onlineSavingsMsg'
Learn More

EXOSC3 Antibody, Alexa Fluor™ 532, Novus Biologicals™

Product Code. 18400178 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18400178 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18400178 Supplier Novus Biologicals Supplier No. NBP222261AF532

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

EXOSC3 Polyclonal antibody specifically detects EXOSC3 in Human, Mouse, Chicken samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen EXOSC3
Applications Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 532
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias bA3J10.7, CGI-102, exosome complex exonuclease RRP40, exosome component 3MGC15120, exosome component Rrp40, hRrp-40, hRrp40p, p10Rrp40p, Ribosomal RNA-processing protein 40, RP11-3J10.8, RRP40MGC723
Host Species Rabbit
Immunogen The immunogen this antibody was made to, maps to a region between residue 225 and 275 of human Exosome Component 3 using the numbering given in entry NP_057126.2 (GeneID 51010).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline DNA replication Transcription Translation and Splicing
Primary or Secondary Primary
Gene ID (Entrez) 51010
Target Species Human, Mouse, Chicken
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.