missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ EXOSC10 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA555869
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Immunogen sequence: KVTELEDKFD LLVDANDVIL ERVGILLDEA SGVNKNQQPV LPAGLQVPKT VVSSWNRKAA EYGKKAKSET FRLLHA Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 93%.
About 50% of patients with polymyositis/scleroderma (PM-Scl) overlap syndrome are reported to have autoantibodies to a nuclear/nucleolar particle termed PM-Scl. Exosome component 10 (EXOSC10), also named autoantigen PM/Scl-2, is the 100 kDa antigen component of PM-Scl and is recognized by most sera of PM-Scl patients. EXOSC10 is strongly enriched in the nucleolus and a small amount has been found in cytoplasm supporting the existence of a nucleolar RNA exosome complex form. As a putative catalytic component ofthe RNA exosome complex which has 3'->5' exoribonuclease activity, EXOSC10 participates in a multitude of cellular RNA processing and degradation events.
Spécification
| EXOSC10 | |
| Polyclonal | |
| Unconjugated | |
| EXOSC10 | |
| Autoantigen PM/Scl 2; Autoantigen PM/Scl 2 homolog; autoantigen PM-SCL; Exosc10; Exosome component 10; P100 polymyositis-scleroderma overlap syndrome-associated autoantigen; p2; p3; p4; PM/Scl-100; PMSCL; PM-Scl; PMSCL2; Polymyositis/scleroderma autoantigen 100 kDa; polymyositis/scleroderma autoantigen 2; Polymyositis/scleroderma autoantigen 2 homolog; RRP6; Rrp6p | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 5394 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.10 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q01780 | |
| EXOSC10 | |
| Recombinant protein corresponding to Human PM-Scl. Recombinant protein control fragment (Product #RP-94454). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu