missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EXOSC10 Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
£119.00 - £295.00
Specifications
| Antigen | EXOSC10 |
|---|---|
| Clone | 1D6G9 |
| Concentration | 1000 μg/mL |
| Applications | Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30237466
|
Proteintech
68315-1-IG-150UL |
150 μL |
£349.00 £295.00 / 150µL Save £54.00 15% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30233262
|
Proteintech
68315-1-IG-20UL |
20 μL |
£135.00 £119.00 / 20µL Save £16.00 11% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The EXOSC10 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human EXOSC10. This antibody recognizes human antigen. The EXOSC10 antibody has been validated for the following applications: WB, IF/ICC, IP, ELISA analysis.Specifications
| EXOSC10 | |
| 1000 μg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| 5394 | |
| Recombinant Protein | |
| Primary | |
| Store at -20°C. Stable for one year after shipment. |
| 1D6G9 | |
| Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, ELISA | |
| Unconjugated | |
| Mouse | |
| Human | |
| PM-Scl; PM/Scl-100; PMSCL; PMSCL2; RRP6; Rrp6p; p2; p3; p4 | |
| IgG1 | |
| Protein G purification | |
| Monoclonal Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title