missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ ExoSAP-IT™ PCR Product Cleanup Reagent

Product Code. 15839906
Change view
Click to view available options
No. of Reactions:
100 Reactions
2000 Reactions
500 Reactions
5000 Reactions
Unit Size:
1mL
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Reactions Unit Size
15839906 5000 Reactions Each
15829906 2000 Reactions Each
14880652 500 Reactions 1mL
12664537 100 Reactions Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 15839906 Supplier Applied Biosystems™ Supplier No. 78205.10.ML

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

ExoSAP-IT™ PCR Product Clean-up Reagent is used for enzymatic clean-up of amplified PCR product. It hydrolyses excess primers and nucleotides in a single step.

  • Remove excess primers and unincorporated nucleotides enzymatically
  • Prepare DNA sequencing samples with one pipetting step
  • Conserve precious PCR products with up to 100% recovery
  • Minimize column clean-up and sample loss
TRUSTED_SUSTAINABILITY

Especificaciones

Content And Storage Shipped on dry ice. Store at -20°C.
Description ExoSAP-IT™ PCR Product Clean-up Reagent, 5000 Reactions, Recombinant, Dry Ice, DNA Sequencing or Single Nucleotide Polymorphism (SNP) Analysis, Use with PCR Purification
Sample Type PCR Products
For Use With (Application) PCR Purification
No. of Reactions 5000 Reactions
Product Line ExoSAP-IT
Protein Form Recombinant
Quantity 10 mL
Research Category Genetic Diseases
Shipping Condition Dry Ice
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.