missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ ExoSAP-IT™ PCR Product Cleanup Reagent
Description
- Remove excess primers and unincorporated nucleotides enzymatically
- Prepare DNA sequencing samples with one pipetting step
- Conserve precious PCR products with up to 100% recovery
- Minimize column clean-up and sample loss
Specifications
Specifications
| Content And Storage | Shipped on dry ice. Store at -20°C. |
| Description | ExoSAP-IT™ PCR Product Clean-up Reagent, 2000 Reactions, Recombinant, Dry Ice, DNA Sequencing or Single Nucleotide Polymorphism (SNP) Analysis, Use with PCR Purification |
| Sample Type | PCR Products |
| For Use With (Application) | PCR Purification |
| No. of Reactions | 2000 Reactions |
| Product Line | ExoSAP-IT |
| Protein Form | Recombinant |
| Quantity | 4 x 1 mL |
| Research Category | Genetic Diseases |
| Shipping Condition | Dry Ice |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?