missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ ExoSAP-IT™ PCR Product Cleanup Reagent
Click to view available options
No. of Reactions:
100 Reactions
2000 Reactions
500 Reactions
5000 Reactions
Unit Size:
1mL
Each
Description
- Remove excess primers and unincorporated nucleotides enzymatically
- Prepare DNA sequencing samples with one pipetting step
- Conserve precious PCR products with up to 100% recovery
- Minimize column clean-up and sample loss
Specifications
Specifications
| Content And Storage | Shipped on dry ice. Store at -20°C. |
| Description | ExoSAP-IT™ PCR Product Clean-up Reagent, 500 Reactions, Recombinant, Dry Ice, DNA Sequencing or Single Nucleotide Polymorphism (SNP) Analysis, Use with PCR Purification |
| Sample Type | PCR Products |
| For Use With (Application) | PCR Purification |
| No. of Reactions | 500 Reactions |
| Product Line | ExoSAP-IT™ |
| Protein Form | Recombinant |
| Quantity | 1 mL |
| Research Category | Genetic Diseases |
| Shipping Condition | Dry Ice |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction