missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ ExoSAP-IT Express PCR Product Cleanup with Tracking Dye
Click to view available options
No. of Reactions:
1920 reactions
500 Reactions
Unit Size:
500 reactions
Each
Description
- 5 minute protocol: Fastest turnaround time to results
- One-tube, one-step PCR clean-up: Add ExoSAP-IT Express directly to PCR product
- Novel enzyme technology: Irreversibly inactivated in just one minute at 80°C
- Conserve PCR samples: 100% recovery of PCR products, regardless of amplicon length
- Eliminate spin columns or beads: Significantly decreases time and errors while increasing yield
- Compatible: No interference in downstream applications
- Scalable: Treat a wide range of volumes
- Multiple formats: Single tube, 8 tube strip, and 96-well plate formats offer flexibility from manual pipetting to automated workflows
- Go green: Generate less waste with a single-tube solution to PCR clean-up
- Optional tracking dye: Never lose track of reagent addition during pipetting
Specifications
Specifications
| Content And Storage | Shipped on dry ice. Store at -20°C. |
| Format | 8 Tube Strip, 96-well Plate, Tube |
| For Use With (Application) | PCR Purification |
| No. of Reactions | 1920 reactions |
| Product Line | ExoSAP-IT™ |
| Protein Form | Recombinant |
| Quantity | 1,920 reactions (96-well plate, 40 μL per we |
| Research Category | Genetic Diseases |
| Shipping Condition | Dry Ice |
| Test Time | 5 min. |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction