missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ ExoSAP-IT Express PCR Product Cleanup with Tracking Dye

Product Code. 15889916
Click to view available options
No. of Reactions:
1920 reactions
500 Reactions
Unit Size:
500 reactions
Each
This item is not returnable. View return policy

Product Code. 15889916

Brand: Applied Biosystems™ A35004

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Our fastest PCR clean-up method - just 5 minutes to superior sequencing results. Now available with optional tracking dye. It is necessary to remove excess primers and unincorporated nucleotides from the PCR reaction prior to sequencing.

  • 5 minute protocol: Fastest turnaround time to results
  • One-tube, one-step PCR clean-up: Add ExoSAP-IT Express directly to PCR product
  • Novel enzyme technology: Irreversibly inactivated in just one minute at 80°C
  • Conserve PCR samples: 100% recovery of PCR products, regardless of amplicon length
  • Eliminate spin columns or beads: Significantly decreases time and errors while increasing yield
  • Compatible: No interference in downstream applications
  • Scalable: Treat a wide range of volumes
  • Multiple formats: Single tube, 8 tube strip, and 96-well plate formats offer flexibility from manual pipetting to automated workflows
  • Go green: Generate less waste with a single-tube solution to PCR clean-up
  • Optional tracking dye: Never lose track of reagent addition during pipetting
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Shipped on dry ice. Store at -20°C.
For Use With (Application) Sequencing, PCR Purification
No. of Reactions 500 Reactions
Product Line ExoSAP-IT™
Product Type PCR Product Cleanup
Protein Form Recombinant
Quantity 500 reactions
Research Category Genetic Diseases
Shipping Condition Dry Ice

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.