missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ ExoSAP-IT™ Express PCR Product Cleanup Reagent

Product Code. 15553677
Change view
Click to view available options
No. of Reactions:
100
2000
480 reactions, 8-tube strip
500
5000
Unit Size:
100 reactions
2000 reactions
480 reactions
500 reactions
5000 reactions
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Reactions unitSize
15553677 5000 5000 reactions
15543677 500 500 reactions
15573677 2000 2000 reactions
15563677 100 100 reactions
15533677 480 reactions, 8-tube strip 480 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 15553677 Supplier Applied Biosystems™ Supplier No. 75001.10.ML

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Removes excess primers and dNTPs. Does not interfere with downstream applications.

  • Mixture of two hydrolytic enzymes, a modified Exonuclease I and Shrimp Alkaline Phosphatase (SAP), in a specially formulated buffer
  • One-tube, one-step PCR cleanup
  • Five-minute protocol
  • Single tube, 8-tube strip, and 96-well plate formats offer flexibility from manual pipetting to automated workflows
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage -20°C
No. of Reactions 5000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.