missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ ExoSAP-IT™ Express PCR Product Cleanup Reagent

Product Code. 15533677
Click to view available options
No. of Reactions:
100
2000
480 reactions, 8-tube strip
500
5000
Unit Size:
100 reactions
2000 reactions
480 reactions
500 reactions
5000 reactions
This item is not returnable. View return policy

Product Code. 15533677

Brand: Applied Biosystems™ 75001.1.EA

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Removes excess primers and dNTPs. Does not interfere with downstream applications.

  • Mixture of two hydrolytic enzymes, a modified Exonuclease I and Shrimp Alkaline Phosphatase (SAP), in a specially formulated buffer
  • One-tube, one-step PCR cleanup
  • Five-minute protocol
  • Single tube, 8-tube strip, and 96-well plate formats offer flexibility from manual pipetting to automated workflows
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage -20°C
No. of Reactions 480 reactions, 8-tube strip
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.