Learn More
Applied Biosystems™ Exonuclease I, high concentration, (20 units/μL)
Product Code. 15859896
Click to view available options
Quantity:
5 units
Unit Size:
Each
Product Code. 15859896
Brand: Applied Biosystems™ 720735KU
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Exonuclease I can be used to measure the endonucleolytic cleavage of covalently closed circular single-stranded DNA reacted with an endonuclease of interest.
Exonuclease I hydrolyzes single-stranded DNA in the 3'→5' direction, releasing. 5'-mononucleotides and leaving the terminal 5'-dinucleotide intact. Hydrolysis is processive and cannot proceed if the 3' terminus is phosphorylated.
Order Info
Each
TRUSTED_SUSTAINABILITY
Specifications
Concentration 20 U/μL
Enzyme Exonuclease
Compatible Buffer Storage Buffer, Glycine Buffer
Quantity 5 units
Product Type Exonuclease I