missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ Exonuclease I, High Conc.: 20U/μL

Product Code. 15523677
Change view
Click to view available options
Quantity:
5000 U
Unit Size:
50000 units
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15523677 5000 U 50000 units
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15523677 Supplier Applied Biosystems™ Supplier No. 720735KU

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Useful in preparing PCR products for applications involving sequencing or labeling methods

Exonuclease I hydrolyzes single-stranded DNA in the 3'→5' direction, releasing 5' mononucleotides and leaving the terminal 5' dinucleotide intact.

TRUSTED_SUSTAINABILITY

Specifications

Concentration 20 U/μL
Purity >95% by SDS-PAGE
Storage Buffer 20mM Tris-HCl (pH 7.5), 5mM 2-mercaptoethanol, 0.5mM EDTA, 50% glycerol
Source E. coli
Quantity 5000 U
Product Type Exonuclease I
Molecular Weight (g/mol) 55kDa
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.