missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EXOG, Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit Polyclonal Antibody
Brand: Abnova H00009941-D01P.100ug
This item is not returnable.
View return policy
Description
Sequence: MAIKSIASRLRGSRRFLSGFVAGAVVGAAGAGLAALQFFRSQGAEGALTGKQPDGSAEKAVLEQFGFPLTGTEARCYTNHALSYDQAKRVPRWVLEHISKSKIMGDADRKHCKFKPDPNIPPTFSAFNEDYVGSGWSRGHMAPAGNNKFSSKAMAETFYLSNIVPQDFDNNSGYWNRIEMYCRELTERFEDVWVVSGPLTLPQTRGDGKKIVSYQVIGEDNVAVPSHLYKVILARRSSVSTEPLALGAFVVPNEAIGFQPQLTEFQVSLQDLEKLSGLVFFPHLDRTSDIRNICSVDTCKLLDFQEFTLYLSTRKIEGARSVLRLEKIMENLKNAEIEPDDYFMSRYEKKLEELKAKEQSGTQIRKPSSpecifications
| EXOG | |
| Polyclonal | |
| In 1x PBS, pH 7.4 | |
| NM_005107 | |
| EXOG | |
| EXOG (NP_005098.1, 1 a.a. ∼ 368 a.a) full-length human protein. | |
| RUO | |
| 9941 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Purified |
| Western Blot | |
| Unconjugated | |
| endo/exonuclease (5'-3'), endonuclease G-like | |
| ENDOGL1/ENDOGL2/ENGL/ENGL-B/ENGL-a/MGC125944/MGC125945 | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction