Learn More
EXOC7 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA554618
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: DDMLDVETDA YIHCVSAFVK LAQSEYQLLA DIIPEHHQKK TFDSLIQDAL DGLMLEGENI VSAARKAIVR HDFSTVLTVF PILRHLKQTK PEFDQVLQGT AASTKNKLPG LITSMETIGA KA Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 97%.
The protein encoded by this gene is a component of the exocyst complex. The exocyst complex plays a critical role in vesicular trafficking and the secretory pathway by targeting post-Golgi vesicles to the plasma membrane. The encoded protein is required for assembly of the exocyst complex and docking of the complex to the plasma membrane. The encoded protein may also play a role in pre-mRNA splicing through interactions with pre-mRNA-processing factor 19. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene, and a pseudogene of this gene is located on the long arm of chromosome 4.Specifications
EXOC7 | |
Polyclonal | |
Unconjugated | |
EXOC7 | |
2-5-3p; EX070; EXO70; Exo70p; EXOC1; Exoc7; Exocyst complex component 7; exocyst complex component Exo70; exocyst component protein 70 kDa homolog; HGNC:23214; KIAA1067; rExo70; sec70; YJL085W | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23265 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9UPT5 | |
EXOC7 | |
Recombinant protein corresponding to Human EXOC7 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only