Learn More
Invitrogen™ EXOC5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5109698
Description
Immunogen sequence: SIFISYLENY IEVETGYLKS RSAMILQRYY DSKNHQKRSI GTGGIQDLKE RIRQRTNLPL GPSIDTHGET FLSQEVVVNL LQETKQAFER CHRLSDPSDL PRNAFRIFTI LVEFLCIEHI DYALETGLAG IPSSDSRNAN LYFLDVVQQA NTIFHLFDKQ FNDHLMPLIS SSPKLSECLQ KKKEIIEQME MKLDTGIDRT LNCMIGQMKH ILAAEQKKTD FKPEDENNVL IQYTNACVKV CAYVRKQVEK IKNSMDGKNV DTVLMELGVR F.
The protein encoded by this gene is a component of the exocyst complex, a multiple protein complex essential for targeting exocytic vesicles to specific docking sites on the plasma membrane. Though best characterized in yeast, the component proteins and functions of exocyst complex have been demonstrated to be highly conserved in higher eukaryotes. At least eight components of the exocyst complex, including this protein, are found to interact with the actin cytoskeletal remodeling and vesicle transport machinery. The complex is also essential for the biogenesis of epithelial cell surface polarity.
Specifications
| EXOC5 | |
| Polyclonal | |
| Unconjugated | |
| Exoc5 | |
| 71 kDa component of rsec6/8 secretory complex; AI447711; AI448003; DKFZp666H126; Exoc5; Exocyst complex component 5; exocyst complex component Sec10; Gm76; hSec10; LOC531907 protein; p71; p71 component of rsec6/8 secretory complex; PRO1912; SEC10; SEC10 protein; Sec10l1; SEC10-like 1; SEC10P; zgc:175220 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 105504, 10640, 60627 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 2.03 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O00471, P97878, Q3TPX4 | |
| Exoc5 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 340-610 of human EXOC5 (NP_006535.1). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.