missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EXOC3, Mouse anti-Human, Clone: 4A7, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00011336-M01.100ug
This item is not returnable.
View return policy
Description
Sequence: SGFGEDVDGYCDTIVAVAEVIKLTDPSLLYLEVSTLVSKYPDIRDDHIGALLAVRGDASRDMKQTIMETLEQGPAQASPSYVPLFKDIVVPSLNVAKLLKSpecifications
| EXOC3 | |
| Monoclonal | |
| Unconjugated | |
| exocyst complex component 3 | |
| SEC6/SEC6L1/Sec6p | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Western Blot | |
| 4A7 | |
| In 1x PBS, pH 7.4 | |
| NM_007277 | |
| EXOC3 | |
| EXOC3 (NP_009208, 646 a.a. ∼ 745 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 11336 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction