missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EXOC2 Antibody, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-33440-20ul
This item is not returnable.
View return policy
Description
EXOC2 Monoclonal antibody specifically detects EXOC2 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| EXOC2 | |
| Monoclonal | |
| Western Blot 1:200 - 1:500, ELISA Recommended starting concentration is 1 μg/mL | |
| exocyst complex component 2, Exocyst complex component Sec5, FLJ11026, SEC5, SEC5L1, SEC5-like 1, SEC5-like 1 (S. cerevisiae), Sec5p | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-100 of human EXOC2 (NP_060773.3).,, Sequence:, MSRSRQPPLVTGISPNEGIPWTKVTIRGENLGTGPTDLIGLTICGHNCLLTAEWMSASKIVCRVGQAKNDKGDIIVTTKSGGRGTSTVSFKLLKPEKIGI | |
| 20 μL | |
| Signal Transduction | |
| 55770 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction