missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EXOC2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£145.00 - £417.00
Specifications
| Antigen | EXOC2 |
|---|---|
| Dilution | Western Blot 1:500 - 1:2000, ELISA, Immunocytochemistry/Immunofluorescence 1:50 - 1:200, Immunoprecipitation 0.5ug - 4ug antibody for 200ug - 400ug extracts of whole cells |
| Applications | ELISA, Immunoprecipitation, Western Blot, Immunocytochemistry/Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30231266
|
Novus Biologicals
NBP3-37953-100ul |
100 μL |
£417.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30228546
|
Novus Biologicals
NBP3-37953-20ul |
20 μL |
£145.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
EXOC2 Polyclonal antibody specifically detects EXOC2 in Human samples. It is validated for ELISA,Immunoprecipitation,Western Blot,Immunocytochemistry/ImmunofluorescenceSpecifications
| EXOC2 | |
| ELISA, Immunoprecipitation, Western Blot, Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Signal Transduction | |
| PBS (pH 7.3), 50% glycerol | |
| 55770 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500 - 1:2000, ELISA, Immunocytochemistry/Immunofluorescence 1:50 - 1:200, Immunoprecipitation 0.5ug - 4ug antibody for 200ug - 400ug extracts of whole cells | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| exocyst complex component 2, Exocyst complex component Sec5, FLJ11026, SEC5, SEC5L1, SEC5-like 1, SEC5-like 1 (S. cerevisiae), Sec5p | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-220 of human EXOC2 (NP_060773.3).,, Sequence:, MSRSRQPPLVTGISPNEGIPWTKVTIRGENLGTGPTDLIGLTICGHNCLLTAEWMSASKIVCRVGQAKNDKGDIIVTTKSGGRGTSTVSFKLLKPEKIGILDQSAVWVDEMNYYDMRTDRNKGIPPLSLRPANPLGIEIEKSKFSQKDLEMLFHGMSADFTSENFSAAWYLIENHSNTSFEQLKMAVTNLKRQANKKSEGSLAYVKGGLSTFFEAQDALS | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title