missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ EXO1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA584141
This item is not returnable.
View return policy
Description
Immunogen sequence: LDETAVTDKE NNLHESEYGD QEGKRLVDTD VARNSSDDIP NNHIPGDHIP DKATVFTDEE SYSFKSSKFT RTISPPTLGT LRSCFSWSGG LGDFSRTPSP SPSTALQQFR RKSDSPTSLP ENNMSDVSQL K Highest antigen sequence indentity to the following orthologs: Rat - 39%, Mouse - 46%.
Substrate-recognition component of the SCF (SKP1-CUL1-F-box protein)-type E3 ubiquitin ligase complex. Required for muscle atrophy following denervation.
Specifications
| EXO1 | |
| Polyclonal | |
| Unconjugated | |
| EXO1 | |
| 5730442G03Rik; Exo1; EXOI; Exonuclease 1; exonuclease I; HEX1; hExo1; hExoI; mExo1; Msa; rad2 nuclease family member, homolog of S. cerevisiae exonuclease 1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 9156 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9UQ84 | |
| EXO1 | |
| Recombinant protein corresponding to Human EXO1. Recombinant protein control fragment (Product #RP-104961). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction