Learn More
EXO1 Mouse, anti-Human, Clone: 2D11D1, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 68536-1-IG-20UL
 View more versions of this product
Product Code. 18050486
  • £135.00 / 20µL
Description
The EXO1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human EXO1. This antibody recognizes human antigen. The EXO1 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
EXO1
2D11D1
EXO1, EXOI, exonuclease 1, Exonuclease I, HEX1, hExo1, hExoI
20 μL
9156
Store at -20°C. Stable for one year after shipment.
Liquid
Western Blot, ELISA
1000 μg/mL
Mouse
Primary
Human
150 kDa
IgG1