missing translation for 'onlineSavingsMsg'
Learn More

Exo1, MedChemExpress

Product Code. 16430967
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16430967 50 mg 50mg
16415516 100 mg 100mg
16414678 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16430967 Supplier Medchem Express Supplier No. HY112670
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Exo1 is a chemical inhibitor of the exocytic pathway.

Exo1 is a chemical inhibitor of the exocytic pathway.

Specifications

Chemical Name or Material Exo1
CAS 75541-83-2
Color White
Purity Grade Notes Research
Molecular Formula C15H12FNO3
Quantity 50 mg
Solubility Information DMSO : 25 mg/mL (91.49 mM; Need ultrasonic)
SMILES O=C(OC)C1=CC=CC=C1NC(C2=CC=C(F)C=C2)=O
Molecular Weight (g/mol) 273.26
Formula Weight 273.26
Percent Purity 99.81%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.