missing translation for 'onlineSavingsMsg'
Learn More
Learn More
exo-IWR 1, Tocris Bioscience™
Negative control for endo-IWR 1 (Cat. No. 3532)
Chemical Identifiers
CAS | 1127442-87-8 |
---|---|
Molecular Formula | C25H19N3O3 |
Molecular Weight (g/mol) | 409.445 |
InChI Key | ZGSXEXBYLJIOGF-BTYSMDAFSA-N |
Synonym | iwr-1-exo, exo-iwr 1, 4-3ar,4r,7s,7as-1,3-dioxo-1,3,3a,4,7,7a-hexahydro-2h-4,7-methanoisoindol-2-yl-n-quinolin-8-yl benzamide, exo-iwr-1, 4-3ar,4r,7s,7as-rel-1,3,3a,4,7,7a-hexahydro-1,3-dioxo-4,7-methano-2h-isoindol-2-yl-n-8-quinolinylbenzamide, rel-4-3ar,4r,7s,7as-1,3,3a,4,7,7a-hexahydro-1,3-dioxo-4,7-methano-2h-isoindol-2-yl-n-8-quinolinylbenzamide |
PubChem CID | 1163034 |
ChEBI | CHEBI:62883 |
SMILES | C1C2C=CC1C3C2C(=O)N(C3=O)C4=CC=C(C=C4)C(=O)NC5=CC=CC6=C5N=CC=C6 |
Description
Negative control for endo-IWR 1 25-fold less active than endo-IWR 1; exhibits decreased activity against the Wnt/β-catenin pathway.Active Analog also available.
Chemical Identifiers
1127442-87-8 | |
409.445 | |
iwr-1-exo, exo-iwr 1, 4-3ar,4r,7s,7as-1,3-dioxo-1,3,3a,4,7,7a-hexahydro-2h-4,7-methanoisoindol-2-yl-n-quinolin-8-yl benzamide, exo-iwr-1, 4-3ar,4r,7s,7as-rel-1,3,3a,4,7,7a-hexahydro-1,3-dioxo-4,7-methano-2h-isoindol-2-yl-n-8-quinolinylbenzamide, rel-4-3ar,4r,7s,7as-1,3,3a,4,7,7a-hexahydro-1,3-dioxo-4,7-methano-2h-isoindol-2-yl-n-8-quinolinylbenzamide | |
CHEBI:62883 |
C25H19N3O3 | |
ZGSXEXBYLJIOGF-BTYSMDAFSA-N | |
1163034 | |
C1C2C=CC1C3C2C(=O)N(C3=O)C4=CC=C(C=C4)C(=O)NC5=CC=CC6=C5N=CC=C6 |
Specifications
1127442-87-8 | |
iwr-1-exo, exo-iwr 1, 4-3ar,4r,7s,7as-1,3-dioxo-1,3,3a,4,7,7a-hexahydro-2h-4,7-methanoisoindol-2-yl-n-quinolin-8-yl benzamide, exo-iwr-1, 4-3ar,4r,7s,7as-rel-1,3,3a,4,7,7a-hexahydro-1,3-dioxo-4,7-methano-2h-isoindol-2-yl-n-8-quinolinylbenzamide, rel-4-3ar,4r,7s,7as-1,3,3a,4,7,7a-hexahydro-1,3-dioxo-4,7-methano-2h-isoindol-2-yl-n-8-quinolinylbenzamide | |
C1C2C=CC1C3C2C(=O)N(C3=O)C4=CC=C(C=C4)C(=O)NC5=CC=CC6=C5N=CC=C6 | |
1163034 | |
409.44 | |
exo-IWR 1 |
C25H19N3O3 | |
ZGSXEXBYLJIOGF-BTYSMDAFSA-N | |
409.445 | |
CHEBI:62883 | |
>99% |
Safety and Handling
Recommended Storage : Store at Room Temperature