missing translation for 'onlineSavingsMsg'
Learn More

exo-7-Oxabicyclo[2.2.1]heptane-2,3-dicarboxylic anhydride, 98%

Product Code. 11367905
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11367905 5 g 5g
11377905 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11367905 Supplier Thermo Scientific Alfa Aesar Supplier No. L02947.06
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 29745-04-8 | C8H8O4 | 168.148 g/mol

exo-7-Oxabicyclo[2.2.1]heptane-2,3-dicarboxylic anhydride acts as a potential antitumor agent. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
exo-7-Oxabicyclo[2.2.1]heptane-2,3-dicarboxylic anhydride acts as a potential antitumor agent. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.

Solubility
Hydrolyzes in water.

Notes
Moisture Sensitive. Store away from moisture and oxidizing agents. Store under dry inert gas. Incompatible with Strong acids, Strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 29745-04-8
Molecular Formula C8H8O4
Molecular Weight (g/mol) 168.148
MDL Number MFCD00213361
InChI Key JAABVEXCGCXWRR-GUCUJZIJSA-N
Synonym exo-3,6-Epoxyhexahydrophthalic anhydride; Norcantharadin
PubChem CID 12251906
SMILES C1CC2C3C(C1O2)C(=O)OC3=O

Specifications

Melting Point 111°C to 116°C
Quantity 5 g
Beilstein 84287
Sensitivity Moisture sensitive
Solubility Information Hydrolyzes in water.
Formula Weight 168.15
Percent Purity ≥98%
Chemical Name or Material exo-7-Oxabicyclo[2.2.1]heptane-2,3-dicarboxylic anhydride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.