missing translation for 'onlineSavingsMsg'
Learn More

exo-7-Oxabicyclo[2.2.1]hept-5-ene-2,3-dicarboxylic anhydride, 98+%

Product Code. 11332246
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11332246

Brand: Thermo Scientific Alfa Aesar L03620.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 6118-51-0 | C8H6O4 | 166.13 g/mol

Intermediate in a useful synthesis of 4,4-dialkyl-2-butenolides, based on reaction with two moles of Grignard reagent, followed by cycloreversion (elimination of furan) to release the double bond.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Intermediate in a useful synthesis of 4,4-dialkyl-2-butenolides, based on reaction with two moles of Grignard reagent, followed by cycloreversion (elimination of furan) to release the double bond.

Solubility
Hydrolyzes in water.

Notes
Moisture sensitive. Store away from strong oxidizing agents, water/ moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6118-51-0
Molecular Formula C8H6O4
Molecular Weight (g/mol) 166.13
MDL Number MFCD00151506,MFCD00077442,MFCD00151506,MFCD00151506
InChI Key QQYNRBAAQFZCLF-UHFFFAOYNA-N
Synonym exo-3,6-epoxy-1,2,3,6-tetrahydrophthalic anhydride
PubChem CID 124671625
IUPAC Name 4,10-dioxatricyclo[5.2.1.0²,⁶]dec-8-ene-3,5-dione
SMILES O=C1OC(=O)C2C3OC(C=C3)C12

Specifications

Melting Point 118°C to 119°C
Quantity 5 g
Beilstein 10355
Sensitivity Moisture sensitive
Solubility Information Hydrolyzes in water.
Formula Weight 166.13
Percent Purity ≥98%
Chemical Name or Material exo-7-Oxabicyclo[2.2.1]hept-5-ene-2,3-dicarboxylic anhydride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.