Learn More
SCAT Exhaust Filter, V3.0
GREENER_CHOICE
Product Code. 17288870 Shop All SCAT Europe Products
Change view
Click to view available options
Size:
L
Large
M
S
Includes:
Multi-Pk. (Service Life 2 x 12 Months)
Multi-Pk. (Service Life 2 x 6 Months)
Multi-Pk. (Service Life 4 x 3 Months)
Single Pk. (Service Life 12 Months)
Single Pk. (Service Life 3 Months)
Single Pk. (Service Life 6 Months)
Display Type:
Indicator
Label
Unit Size:
Each
Pack of 2
Pack of 4
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Includes Display Type unitSize
17288870 L Multi-Pk. (Service Life 2 x 12 Months) Indicator Pack of 2
17238870 S Single Pk. (Service Life 3 Months) Indicator Each
17258870 S Multi-Pk. (Service Life 4 x 3 Months) Indicator Pack of 4
17218870 M Single Pk. (Service Life 6 Months) Label Each
17248870 M Single Pk. (Service Life 6 Months) Indicator Each
17278870 M Multi-Pk. (Service Life 2 x 6 Months) Label Pack of 2
17268870 M Multi-Pk. (Service Life 2 x 6 Months) Indicator Pack of 2
17228870 L Single Pk. (Service Life 12 Months) Indicator Each
17926970 Large Single Pk. (Service Life 12 Months) Label Each
17936970 Large Multi-Pk. (Service Life 2 x 12 Months) Label Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Product Code. 17288870 Supplier SCAT Supplier No. 490986
Only null left
Add to basket
Edge
Add to basket
The SCAT Exhaust Filter V3.0 for SafetyWasteCaps guarantees over 90% CTC adsorption for maximum air purity due to three integrated activated carbon types. This makes it the most powerful activated carbon filter medium on the HPLC laboratory market.
  • Block off harmful vapours and ensure for safe pressure equalization in the waste containers
  • 3-layer filter media with high performance spectrum for acids and alkalis
  • Optimized for binding HPLC buffer solutions and absorbing organic solvent vapors
  • The natural pore structure of the activated carbon prevents sticking or clogging
  • Precise indication of the next replacement time by means of a time indicator
  • Available in sizes (S/M/L) with different service lives (3/6/12 months)
  • The use of exhaust filters can contribute to the reduction of the air exchange rate
Specifications
Display Type Indicator
For Use With (Equipment) SCAT Safety Waste Caps & Universal Waste Hub JAN
Includes Multi-Pk. (Service Life 2 x 12 Months)
Material Polypropylene
Size L
Thread Size GL14
For Use With (Application) HPLC Systems
Type Exhaust Filter