missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Exendin-4 Antibody (35), Alexa Fluor™ 750, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-05179AF750
This item is not returnable.
View return policy
Description
Exendin-4 Monoclonal specifically detects Exendin-4 in Reptile samples. It is validated for ELISA.
Specifications
| Exendin-4 | |
| Monoclonal | |
| Alexa Fluor 750 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| Mouse | |
| 0.1 mL | |
| Specific for exendin-4. No cross-reactivity with GLP-1, GLP-2 (human) or glucagon coated on ELISA wells. | |
| Store at 4°C in the dark. |
| ELISA | |
| 35 | |
| ELISA | |
| Exendin4 | |
| Synthetic exendin-4 | |
| Primary | |
| Human | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction