missing translation for 'onlineSavingsMsg'
Learn More

Exendin-4 Antibody (24), DyLight 755, Novus Biologicals™

Product Code. 18423977 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18423977 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18423977 Supplier Novus Biologicals Supplier No. NBP223579IR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Exendin-4 Monoclonal antibody specifically detects Exendin-4 in Reptile samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen Exendin-4
Applications ELISA
Classification Monoclonal
Clone 24
Conjugate DyLight 755
Formulation 50mM Sodium Borate
Gene Alias Exendin4
Host Species Mouse
Immunogen Synthetic exendin-4
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Biologically Active Proteins
Primary or Secondary Primary
Target Species Reptile
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.