Learn More
BINDER™ Execution of IQ/OQ
Product Code. 17882445 Shop All BINDER Products
Click to view available options
For Use With (Equipment):
Environmental Walk-in Chambers
Unit Size:
Each
Product Code. 17882445
Brand: BINDER™ DL400200
Execution of IQ/OQ in accordance with qualification folder.
  • Installation Qualification (IQ) confirms that the unit has been properly installed according to customer requirements including documentation. The technician checks that the unit has been installed correctly, as specified in the qualification folder. The qualification folders can be ordered on a unit-specific basis.
  • OQ (Operational Qualification) checks and confirms that the unit is operating properly in an unloaded state. The required tests are available in the qualification folder.
Specifications
For Use With (Equipment) Environmental Walk-in Chambers
Product Type IQ/OQ Service