missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ EXD2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA552218
This item is not returnable.
View return policy
Description
Immunogen sequence: TSCHAISNYY DNHLKQQLAK EFQAPIGSEE GLRLLEDPER RQVRSGARAL LNAESLPTQR KEELLQALRE FYNTDVVTEE MLQEAASLET RISNENYVPH GLKVVQCH Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 92%.
Exonuclease that has both 3'-5' exoribonuclease and exodeoxyribonuclease activities, depending on the divalent metal cation used as cofactor (PubMed:29335528, PubMed:31127291). In presence of Mg(2+), only shows 3'-5' exoribonuclease activity, while it shows both exoribonuclease and exodeoxyribonuclease activities in presence of Mn(2+) (PubMed:29335528, PubMed:31127291). Acts as an exoribonuclease in mitochondrion, possibly by regulating ATP production and mitochondrial translation (PubMed:29335528). Also involved in the response to DNA damage (PubMed:26807646, PubMed:31255466). Acts as 3'-5' exodeoxyribonuclease for double-strand breaks resection and efficient homologous recombination (PubMed:20603073, PubMed:26807646). Plays a key role in controlling the initial steps of chromosomal break repair, it is recruited to chromatin in a damage-dependent manner and functionally interacts with the MRN complex to accelerate resection through its 3'-5' exonuclease activity, which efficiently processes double-stranded DNA substrates containing nicks (PubMed:26807646). Also involved in response to replicative stress: recruited to stalled forks and is required to stabilize and restart stalled replication forks by restraining excessive fork regression, thereby suppressing their degradation (PubMed:31255466). [UniProt].
Specifications
| EXD2 | |
| Polyclonal | |
| Unconjugated | |
| EXD2 | |
| 3'-5' exoribonuclease EXD2; 4930539P14Rik; C14orf114; C85658; EXD2; EXDL2; exonuclease 3'-5' domain containing 2; exonuclease 3'-5' domain-containing protein 2; exonuclease 3''-5'' domain-like 2; exonuclease 3'-5' domain-like 2; exonuclease 3'-5' domain-like-containing protein 2; RGD1311087 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 55218, 97827 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.80 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8VEG4, Q9NVH0 | |
| EXD2 | |
| Recombinant protein corresponding to Human EXD2. Recombinant protein control fragment (Product #RP-89042). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction