missing translation for 'onlineSavingsMsg'
Learn More

EX229, MedChemExpress

Product Code. 16423206
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16423206

Brand: Medchem Express HY112769

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress EX229, a Benzimidazole derivative, is a potent and allosteric activator of AMP-activated protein kinase (AMPK), with Kds of 0.06 μM, 0.06 μM and 0.51 μM for α1β1γ1, α2β1γ1 and α1β2γ1 in biolayer interferometry, respectively.

EX229, a Benzimidazole derivative, is a potent and allosteric activator of AMP-activated protein kinase (AMPK), with Kds of 0.06 μM, 0.06 μM and 0.51 μM for α1β1γ1, α2β1γ1 and α1β2γ1 in biolayer interferometry, respectively.

Specifications

Chemical Name or Material EX229
CAS 1219739-36-2
Color Pink
Purity Grade Notes Research
Molecular Formula C24H18ClN3O3
Quantity 25 mg
Solubility Information DMSO : 13 mg/mL (30.10 mM; Need ultrasonic and warming)
SMILES O=C(O)C1=CC(OC2=NC3=CC(Cl)=C(C4=CC5=C(N(C)C=C5)C=C4)C=C3N2)=CC=C1C
Molecular Weight (g/mol) 431.87
Formula Weight 431.87
Percent Purity 98.05%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.