missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ EWSR1 Monoclonal Antibody (5H7)

Mouse Monoclonal Antibody
£430.00 - £1482.00
Specifications
| Antigen | EWSR1 |
|---|---|
| Clone | 5H7 |
| Concentration | 1 mg/mL |
| Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
| Applications | Immunohistochemistry, Western Blot, Immunocytochemistry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17815157
|
Invitrogen™
MA547354 |
500 μL |
£1482.00
500µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17875197
![]() |
Invitrogen™
MA547453 |
100 μL |
£430.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
EWSR1 Monoclonal Antibody for Western Blot, ICC/IF, IHC
This gene encodes a multifunctional protein that is involved in various cellular processes, including gene expression, cell signaling, and RNA processing and transport. The protein includes an N-terminal transcriptional activation domain and a C-terminal RNA-binding domain. Chromosomal translocations between this gene and various genes encoding transcription factors result in the production of chimeric proteins that are involved in tumorigenesis. These chimeric proteins usually consist of the N-terminal transcriptional activation domain of this protein fused to the C-terminal DNA-binding domain of the transcription factor protein. Mutations in this gene, specifically a ttranslocation, are known to cause Ewing sarcoma as well as neuroectodermal and various other tumors. Alternative splicing of this gene results in multiple transcript variants. Related pseudogenes have been identified on chromosomes 1 and 14.Specifications
| EWSR1 | |
| 1 mg/mL | |
| Immunohistochemistry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Mouse | |
| RUO | |
| PBS with 50% glycerol and 5mM sodium azide | |
| Q01844, Q61545 | |
| 100034010, 14030, 2130, 289752, 609786 | |
| Full length human EWS expressed in and purified from <i>E. coli</i>. | |
| Antibody |
| 5H7 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Monoclonal | |
| Liquid | |
| IgG2b | |
| Human, Mouse, Rat, Canine, Horse | |
| EWSR1 | |
| AC002059.7; bK984G1.4; Ewing sarcoma breakpoint region 1; Ewing sarcoma breakpoint region 1 protein; Ewing sarcoma homolog; Ewings sarcoma EWS-Fli1 (type 1) oncogene; Ews; EWS oncogene; EWS RNA binding protein 1; EWS RNA-binding protein 1; EWS RNA-binding protein variant 6; EWS-FLI1; Ewsh; EWSR1; RNA-binding protein EWS | |
| EWSR1 | |
| Primary | |
| Protein A/G |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title