missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EWSR1 Antibody, DyLight 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB200-183UV
This item is not returnable.
View return policy
Description
EWSR1 Polyclonal antibody specifically detects EWSR1 in Human samples. It is validated for Western Blot
Specifications
| EWSR1 | |
| Polyclonal | |
| Western Blot | |
| bK984G1.4, Ewing sarcoma breakpoint region 1, Ewing sarcoma breakpoint region 1 protein, Ewings sarcoma EWS-Fli1 (type 1) oncogene, EWS oncogene, EWSRNA-binding protein EWS | |
| A synthetic peptide that maps to a region between residues 600 and the C-terminus (residue 656) of human Ewing sarcoma breakpoint region 1 using the numbering given in SwissProt entry Q01844 (GeneID 2130). | |
| 0.1 mL | |
| Cancer, Cellular Markers, Neuroscience, Tumor Suppressors | |
| 2130 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| DyLight 350 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction