missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ERManI Antibody (3C2), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 3 publications
Brand: Novus Biologicals NBP2-13167
This item is not returnable.
View return policy
Description
ERManI Monoclonal specifically detects ERManI in Human, Mouse (Negative) samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Knockout Validated.
Specifications
| ERManI | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot 1:500-1:1000, ELISA 1:1000, Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 1:100, Immunoprecipitation 1:10-1:100, Immunohistochemistry-Paraffin 1:200, Knockout Validated reported in scientific literature (PMID 26205822) | |
| alpha 1,2-mannosidase, EC 3.2.1.113, endoplasmic reticulum alpha-mannosidase 1, endoplasmic reticulum mannosyl-oligosaccharide 1,2-alpha-mannosidase, endoplasmic reticulum mannosyl-oligosaccharide 1,2-alpha-mannosidase 1, ER alpha 1,2-mannosidase, ER alpha-1,2-mannosidase, ER ManI, ER mannosidase 1, ERMan1, MAN1B1, Man9GlcNAc2-specific processing alpha-mannosidase, Man9GlcNAc2-specific-processing alpha-mannosidase, MANA-ER, Mannosidase alpha class 1B member 1, mannosidase, alpha, class 1B, member 1, MRT15, RP11-350O14.2 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| 11253 | |
| Human, Mouse (Negative) | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| 3C2 | |
| Unconjugated | |
| Q9UKM7 | |
| MAN1B1 | |
| Full length human recombinant ERManI [Swiss-Prot# Q9UKM7] | |
| 0.1 mL | |
| Primary | |
| Human. Does not cross react with Mouse. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction