missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Evx2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA559632
This item is not returnable.
View return policy
Description
Immunogen sequence: PLHSALGELP AKGKFEIDTL FNLQHTGSES TVSSEISSAA ESRKKPGHYS Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 86%.
EVX2, a protein related to the Drosophila even-skipped (eve), is expressed in limbs like the neighbouring Hoxd genes. Evx-2 functions like a Hoxd gene.
Specifications
| Evx2 | |
| Polyclonal | |
| Unconjugated | |
| EVX2 | |
| eve, even-skipped homeo box homolog 2; even skipped homeotic gene 2; even skipped homeotic gene 2 homolog; even-skipped homeo box 2 (homolog of Drosophila eve); even-skipped homeobox 2; Evx2; Evx-2; Homeobox even-skipped homolog protein 2; LOW QUALITY PROTEIN: homeobox even-skipped homolog protein 2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 344191 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q03828 | |
| EVX2 | |
| Recombinant protein corresponding to Human Evx2. Recombinant protein control fragment (Product #RP-99916). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction