missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EVPL Antibody, Novus Biologicals™
Description
EVPL Polyclonal antibody specifically detects EVPL in Human samples. It is validated for Immunoprecipitation
Specifications
Specifications
| Antigen | EVPL |
| Applications | Immunoprecipitation |
| Classification | Polyclonal |
| Concentration | 1 mg/ml |
| Conjugate | Unconjugated |
| Dilution | Immunoprecipitation 8 - 15 ug/mg lysate |
| Formulation | Tris-citrate/phosphate pH 7 -8. |
| Gene Alias | 210 KDa Cornified Envelope Precursor Protein, 210 KDa Paraneoplastic Pemphigus Antigen, envoplakin, EVPK, p210 |
| Host Species | Rabbit |
| Immunogen | Recognizes region between residues Between Between 1983 and 2033.. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?