missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ EVOS™ Light Cube, White
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Description
- Contains unique, proprietary LED light cubes
- Image fluorscence samples for specific wavelengthLight engine outputs high intensity over a short light path that delivers superior fluorophore excitation
- Each cube contains an LED, condensing optics, and hard-coated filters
- User-changeable and automatically recognized by system
- Shorter light path that affords superior detection of fluorescent signals
- Continuous illumination that enables consistent results
- 50,000+ hour bulb lifetime that affords lower operating costs
- Adjustable light intensity that helps reduce photobleaching
Specifications
Specifications
| Type | LED Light Cube |
| Product Line | EVOS |
| For Use With (Equipment) | Countess™ II FL Automated Cell Counter, EVOS™ M5000 Imaging System, EVOS™ M7000 Imaging System |
| Includes | Condensing Optics, Hard-Coated Filters, LED |
| Light Source | Metal Halide (Bulb) |
| Quantity | 1 Ea. |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction