Learn More
Thermo Scientific™ EVOS™ Vessel Plate for Large Vessels
This EVOS vessel plate is intended for use on the stage of the EVOS FL Color Imaging System.
Brand:  Thermo Scientific™ AMEPVP01
Code : NEW
Additional Details : Weight : 2.00000kg
Product Code. 15329661
  • £126.00 / Each
Estimated Shipment: 23-08-2024
to see stock.
Description
Description
This EVOS vessel plate is intended for use on the stage of the EVOS FL Color Imaging System. This plate is designed to hold large vessels, for increased precision in sample alignment and is just one of many easy-to-change plates and holders available for this instrument platform. With a snug and secure fit, yet readily switched, these accessories make imaging samples in a range of vessel holders convenient and easy.
Specifications
Specifications
Vessel Plate
Large Vessel
1 Ea.
EVOS
EVOS™ XL Core Imaging System, EVOS™ FL Color Imaging System, EVOS™ XL Imaging System, EVOS™ FL Imaging System, EVOS™ FL Auto Imaging System