missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EVOS™ EVOS™ Vessel Holder, two 25 x 75 mm slides
Click to view available options
Quantity:
1 Holder
Unit Size:
Each
Description
Broad range of holders helps maximize the flexibility of the EVOS and Micromaster™ inverted microscopes.
Specifications
Specifications
| Type | Vessel Holder |
| Product Line | EVOS |
| For Use With (Equipment) | EVOS™ XL Core Imaging System, EVOS™ FL Color Imaging System, EVOS™ XL Imaging System, EVOS™ FL Imaging System, EVOS™ FL Auto Imaging System |
| Format | 2 Slides |
| Holds | 2 25 x 75 mm Microscope Slides, Chamber |
| Quantity | 1 Holder |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction