Learn More
EVOS™ EVOS™ Vessel Holder, Greiner T-75 flask
For flasks, trays and petri dishes
Brand: EVOS™ AMEPVH008
This item is not returnable.
View return policy
Description
Broad range of holders helps maximize the flexibility of the EVOS and Micromaster™ inverted microscopes.
Specifications
Vessel Holder | |
EVOS™ XL Core Imaging System, EVOS™ FL Color Imaging System, EVOS™ XL Imaging System, EVOS™ FL Imaging System, EVOS™ FL Auto Imaging System | |
1 75 cm2 BD/Greiner T-75 flask |
EVOS | |
Greiner T-75 flask | |
1 Holder |
For Research Use Only. Not for use in diagnostic procedures.