Learn More
EVOS™ EVOS™ Vessel Holder, Nunc™/SPL IVF 4-well dish
Product Code. 11967325
Click to view available options
Quantity:
1 Holder
Unit Size:
Each
Product Code. 11967325
Brand: EVOS™ AMEPVH011
For flasks, trays and petri dishes
Broad range of holders helps maximize the flexibility of the EVOS and Micromaster™ inverted microscopes.
TRUSTED_SUSTAINABILITY
Specifications
Type Vessel Holder
Product Line EVOS
For Use With (Equipment) EVOS™ XL Core Imaging System, EVOS™ FL Color Imaging System, EVOS™ XL Imaging System, EVOS™ FL Imaging System, EVOS™ FL Auto Imaging System
Format Nunc 4-well dish for IVF
Holds 1 Nunc/SPL IVF 4-well Dish
Quantity 1 Holder