Learn More
EVOS™ EVOS™ Vessel Holder, one Nunc™ T-25 flask
Click to view available options
Quantity:
1 holder
Unit Size:
Each
Description
Product Features:
- EVOS™ vessel holders allow a perfect fit of your microscope slide, cell culture flask or dish, or micro-well plate to the stage of the EVOS imaging systems for increased precision in sample alignment.
- EVOS stage plates hold vessel holders, thereby expanding the number of vessel holders available for the EVOS systems.
- With a snug and secure fit, yet readily switched, these accessories make imaging samples in a range of vessel holders convenient and easy.
Specifications
Specifications
| Type | Imaging System Vessel Holder |
| Product Line | EVOS |
| Description | Vessel Holder, one Nunc T-25 flask |
| For Use With (Equipment) | EVOS™ FL Auto Imaging System |
| Holds | 1 Nunc T-25 flask |
| Quantity | 1 holder |