missing translation for 'onlineSavingsMsg'
Learn More

EVOS™ EVOS™ Vessel Holder for Automated Stage, with retention clip, one Nunc™ multi-well dish

EVOS™ vessel holders and stage plates fit a wide range of vessels, flasks, plates, dishes, and slides.

Brand:  EVOS™ AMEPVH061

Code : NEW

Additional Details : Weight : 0.50000kg

Product Code. 15461084

  • £153.00 / Each
Estimated Shipment: 05-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Product Features:
  • EVOS™ vessel holders allow a perfect fit of your microscope slide, cell culture flask or dish, or micro-well plate to the stage of the EVOS imaging systems for increased precision in sample alignment.
  • EVOS stage plates hold vessel holders, thereby expanding the number of vessel holders available for the EVOS systems.
  • With a snug and secure fit, yet readily switched, these accessories make imaging samples in a range of vessel holders convenient and easy.
Specifications

Specifications

Imaging System Vessel Holder
Vessel Holder for Automated Stage, one Nunc multi-well dish
1 multi-well plate
Room Temperature
EVOS
EVOS™ FL Auto Imaging System
159 mm x 109 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.