Learn More
EVOS™ EVOS™ Vessel Holder/Adapter for Automated Stage, with retention, multi-well plates
Allows user to utilize different types of vessels for imaging
Brand: EVOS™ AMEPVH022
Code : NEW
Additional Details : Weight : 1.00000kg
Description
- Fits microscope slide, cell culture flask or dish, or micro-well plate to stage of EVOS imaging systems for increased precision in sample alignment
- Holds multi-well plates
- Acts as adapter for standard vessel holders (Cat numbers AMEP-VH001 thru AMEP-VH018)
- Specific for automated stage of EVOS FL Auto Imaging System
- Has lever for added convenience
Specifications
Vessel Holder | |
EVOS™ XL Core Imaging System, EVOS™ FL Color Imaging System, EVOS™ XL Imaging System, EVOS™ FL Imaging System, EVOS™ FL Auto Imaging System | |
Multi-well Plate |
EVOS | |
one, multi-well plate | |
1 Holder |
For Research Use Only. Not for use in diagnostic procedures.